Learn More
Fine Science Tools Colibri Retractors, Stainless Steel, 3 cm
Catalog No. NC9128320
Encompass
Product: Colibri Retractor - Wire/3cm/1.5cm Maximum Spread/ Teeth: Wire/ Teeth Length: 3mm/ Teeth Width: 4mm/ Maximum Spread: 1.5cm/ Alloy: Stainless Steel/ Length: 3cm/ Feature: Slightly Curved/ Unit: Each
Catalog No. NC9128320 Supplier Fine Science Tools Supplier No. 1700003
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Product: Colibri Retractor - Wire/3cm/1.5cm Maximum Spread/ Teeth: Wire/ Teeth Length: 3mm/ Teeth Width: 4mm/ Maximum Spread: 1.5cm/ Alloy: Stainless Steel/ Length: 3cm/ Feature: Slightly Curved/ Unit: Each