Learn More
Cole-Parmer™ Masterflex™ L/S™ Platinum-Cured Silicone High-Performance Precision Tubing, Size L/S 24; Platinum-cured silicone
Ensure optimal performance from your Masterflex™ pump with this silicone pump tubing. Masterflex™ L/S™ Platinum-Cured Silicone High-Performance Precision Tubing provides accurate, repeatable flow. Ideal for Pharmaceutical, biotech, peristaltic pumping and food applications.
Supplier:  Cole-Parmer™ MFLX9641024
Catalog No. MFX9641024
Only null left
Description
Description
  • Manufactured to extremely close tolerances that match L/S™ pump heads, ensuring accurate, repeatable flow and long tubing life
  • Factory-tested and optically inspected to provide the best performance from your peristaltic pump
  • Features a thicker wall compared to our Precision pump tubing, making it the best choice for applications involving pressure, suction lift and viscous fluids
  • Fewer potential leachables
  • Excellent biocompatibility
  • Odorless and nontoxic, with no taste imparted to transported fluids
  • Resistant to fungus growth, weather, ozone, corona and radiation
  • Smoother surface than peroxide-cured silicone for reduced protein binding
  • Excellent heat resistance, low compression set, odorless, nontoxic
  • Translucent
Product Suggestions
Product Suggestions