missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Coils and Core Activity Set

Catalog No. S83616
Change view
Click to view available options
Class Size:
1 Class
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S83616 1 Class
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S83616 Supplier Eisco™ Supplier No. EISPH0850CSET

Coils and Core Activity Set allows students to study how transformers work, as well as several different properties of induced magnetism.

  • Activity kit includes: Plastic base, Laminated iron core with mounting fixtures, 6 Coils, 5 student activity capture sheets, and a manual.
  • Coils are protected in a heat resistant film.
  • Topics covered include Lenz' Law, Faraday's Law, how iron cores increase magnetic field strength, and electromagnetic induction.
  • Can also be used to teach about how transformers used by power companies carry electrical energy.
  • Includes extensive instructions on how to use the apparatus as a demonstration, as well as inquiry based lessons surrounding electromagnetic induction and transformers.

Specifications

Grade 7 to 12
For Use With (Application) Lenz' Law, Faraday's Law, and Electromagnetic Induction
Includes Plastic Base, Laminated Iron Core with Mounting Fixtures, 6 Coils, 5 Student Activity Capture Sheets, and Manual
Class Size 1 Class
Product Type Electricity Kit