Learn More
Synthware™ Rotary Evaporator Condensers
GREENER_CHOICE
For use with rotary evaporators
$4,305.75 - $4,350.14
Specifications
Material Borosilicate Glass
Type Condenser
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
31500820
GREENER_CHOICE
View Documents Promotion Details
Synthware™
C910200
Buchi/Diagonal Style Rotary Evaporator
Each for $4,350.14
Only null left
31500822
GREENER_CHOICE
View Documents Promotion Details
Synthware™
C900200
Upright Style Rotary Evaporator
Each for $4,305.75
Only null left
Description
Description
C900200:
For vertical style of Heifdorfe.
Accessories:
C900001 Inlet feed tube, 19/38 joint 86.55
RC14 Nylon™ cap 1.15
R201402 Replacement hose connection-straight 1.45
C910200:
Double coil condenser for use with Buchi rotary evaporators. For cooling with tap water in a diagonal position. Has a 19/38 top valve with a 35/20 spherical ball joint.
Accessories:
C900001 Inlet feed tube, 19/38 joint 86.55
RC14 Nylon™ cap 1.15
R201402 Replacement hose connection-straight 1.45
Specifications
Specifications
Borosilicate Glass
Condenser