missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Coenzyme Q10, 98%

Catalog No. p-7077702
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC457950010 1 g
AC457950050 5 g
AC457950250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC457950010 Supplier Thermo Scientific Chemicals Supplier No. 457950010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 303-98-0 | C59H90O4 | 863.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 303-98-0
Molecular Formula C59H90O4
Molecular Weight (g/mol) 863.37
MDL Number MFCD00042919
InChI Key ACTIUHUUMQJHFO-UPTCCGCDSA-N
Synonym coenzyme q10, ubidecarenone, ubiquinone 50, coq10, ubiquinone-10, neuquinon, ubiquinone, justquinon, neuquinone, emitolon
PubChem CID 5281915
ChEBI CHEBI:46245
SMILES COC1=C(OC)C(=O)C(C\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CCC=C(C)C)=C(C)C1=O

Specifications

Melting Point 49.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether and ethanol
IUPAC Name 2-[(2E,6E,10E,14E,18E,22E,26E,30E,34E)-3,7,11,15,19,23,27,31,35,39-decamethyltetraconta-2,6,10,14,18,22,26,30,34,38-decaenyl]-5,6-dimethoxy-3-methylcyclohexa-2,5-diene-1,4-dione
Formula Weight 863.37
Percent Purity 98%
Chemical Name or Material Coenzyme Q10
EINECSNumber 206-147-9

RUO – Research Use Only