missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Coenzyme Q10, 98%
Supplier: Thermo Scientific™ 457950050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Coenzyme Q10 | |
| Authentic | |
| C59H90O4 | |
| 5g | |
| Solubility in water: insoluble. Other solubilities: soluble in ether and ethanol | |
| COC1=C(OC)C(=O)C(C\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CCC=C(C)C)=C(C)C1=O | |
| 863.37 | |
| CHEBI:46245 | |
| 98% |
| 303-98-0 | |
| 97.5% min. (HPLC) | |
| MFCD00042919 | |
| coenzyme q10, ubidecarenone, ubiquinone 50, coq10, ubiquinone-10, neuquinon, ubiquinone, justquinon, neuquinone, emitolon | |
| ACTIUHUUMQJHFO-UPTCCGCDSA-N | |
| 2-[(2E,6E,10E,14E,18E,22E,26E,30E,34E)-3,7,11,15,19,23,27,31,35,39-decamethyltetraconta-2,6,10,14,18,22,26,30,34,38-decaen-1-yl]-5,6-dimethoxy-3-methylcyclohexa-2,5-diene-1,4-dione | |
| 5281915 | |
| 863.37 |
Safety and Handling
EINECSNumber : 206-147-9
RUO â Research Use Only