missing translation for 'onlineSavingsMsg'
Learn More

Coenzyme Q-10, 98+%

Component of electron transport chain and in aerobic cellular respiration | CAS: 303-98-0 | C59H90O4 | 863.37 g/mol

Supplier:  Thermo Scientific Chemicals J6513706

 View more versions of this product

Catalog No. AAJ6513706


Only null left
Explore available promotions

Description

Description

It is used as a component of electron transport chain and in aerobic cellular respiration, Coenzyme Q10 can protect human endothelial cells from oxidative damage by modulating the nitric oxide pathway. In addition, Coenzyme Q10 inhibits LDL oxidation, a major process in the development of atherosclerosis. Coenzyme Q10 is known to suppress the down regulation of NOS and up-regulation of NOS2. As a result of its ability to bind Ca2+, Coenzyme Q10 can transport this cation across artificial biomimetic membranes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a component of electron transport chain and in aerobic cellular respiration, Coenzyme Q10 can protect human endothelial cells from oxidative damage by modulating the nitric oxide pathway. In addition, Coenzyme Q10 inhibits LDL oxidation, a major process in the development of atherosclerosis. Coenzyme Q10 is known to suppress the down regulation of NOS and up-regulation of NOS2. As a result of its ability to bind Ca2+, Coenzyme Q10 can transport this cation across artificial biomimetic membranes.

Solubility
Soluble in chloroform.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

Coenzyme Q-10
48°C to 52°C
C59H90O4
5 g
Light sensitive
coenzyme q10, ubidecarenone, ubiquinone 50, coq10, ubiquinone-10, neuquinon, ubiquinone, justquinon, neuquinone, emitolon
ACTIUHUUMQJHFO-UPTCCGCDSA-N
2-[(2E,6E,10E,14E,18E,22E,26E,30E,34E)-3,7,11,15,19,23,27,31,35,39-decamethyltetraconta-2,6,10,14,18,22,26,30,34,38-decaen-1-yl]-5,6-dimethoxy-3-methylcyclohexa-2,5-diene-1,4-dione
5281915
863.36
Powder
303-98-0
Yellow to Orange
MFCD00042919
1900141
14,9843
Soluble in chloroform.
COC1=C(OC)C(=O)C(C\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CCC=C(C)C)=C(C)C1=O
863.37
CHEBI:46245
≥98%

Chemical Identifiers

303-98-0
863.37
ACTIUHUUMQJHFO-UPTCCGCDSA-N
5281915
COC1=C(OC)C(=O)C(C\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CCC=C(C)C)=C(C)C1=O
C59H90O4
MFCD00042919
coenzyme q10, ubidecarenone, ubiquinone 50, coq10, ubiquinone-10, neuquinon, ubiquinone, justquinon, neuquinone, emitolon
CHEBI:46245
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 206-147-9

RTECSNumber : DK3900000

TSCA : Yes

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only