missing translation for 'onlineSavingsMsg'
Learn More

Coenzyme A Trilithium Salt Dihydrate ≥96%, White Crystalline Powder, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0210049390

 View more versions of this product

Catalog No. ICN10049390

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Coenzyme A Trilithium Salt Dihydrate >96%
White
C21H33Li3N7O16P3S
500 mg
QSCBPHBAFBVXRK-UHFFFAOYSA-K
trilithium;[5-(6-aminopurin-9-yl)-4-hydroxy-2-[[[[3-hydroxy-2,2-dimethyl-4-oxo-4-[[3-oxo-3-(2-sulfanylethylamino)propyl]amino]butoxy]-oxidophosphoryl]oxy-oxidophosphoryl]oxymethyl]oxolan-3-yl] hydrogen phosphate
53442190
Crystalline Powder
18439-24-2
≥96%
MFCD00075848
coenzyme a, trilithium salt
[Li+].[Li+].[Li+].CC(C)(COP(=O)([O-])OP(=O)([O-])OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)[O-])C(C(=O)NCCC(=O)NCCS)O
785.329
821.4

Chemical Identifiers

18439-24-2
785.329
QSCBPHBAFBVXRK-UHFFFAOYSA-K
53442190
[Li+].[Li+].[Li+].CC(C)(COP(=O)([O-])OP(=O)([O-])OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)[O-])C(C(=O)NCCC(=O)NCCS)O
C21H33Li3N7O16P3S
MFCD00075848
coenzyme a, trilithium salt
trilithium;[5-(6-aminopurin-9-yl)-4-hydroxy-2-[[[[3-hydroxy-2,2-dimethyl-4-oxo-4-[[3-oxo-3-(2-sulfanylethylamino)propyl]amino]butoxy]-oxidophosphoryl]oxy-oxidophosphoryl]oxymethyl]oxolan-3-yl] hydrogen phosphate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20 °C.

SDS
Documents

Documents

Promotions

Promotions