missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coenzyme A Trilithium Salt Dihydrate ≥96%, White Crystalline Powder, MP Biomedicals™
Supplier: MP Biomedicals Inc 0210049325
Specifications
| Coenzyme A Trilithium Salt Dihydrate >96% | |
| White | |
| C21H33Li3N7O16P3S | |
| 25 mg | |
| QSCBPHBAFBVXRK-UHFFFAOYSA-K | |
| trilithium;[5-(6-aminopurin-9-yl)-4-hydroxy-2-[[[[3-hydroxy-2,2-dimethyl-4-oxo-4-[[3-oxo-3-(2-sulfanylethylamino)propyl]amino]butoxy]-oxidophosphoryl]oxy-oxidophosphoryl]oxymethyl]oxolan-3-yl] hydrogen phosphate | |
| 53442190 | |
| Crystalline Powder |
| 18439-24-2 | |
| ≥96% | |
| MFCD00075848 | |
| coenzyme a, trilithium salt | |
| [Li+].[Li+].[Li+].CC(C)(COP(=O)([O-])OP(=O)([O-])OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)[O-])C(C(=O)NCCC(=O)NCCS)O | |
| 785.329 | |
| 821.4 |
Chemical Identifiers
| 18439-24-2 | |
| 785.329 | |
| QSCBPHBAFBVXRK-UHFFFAOYSA-K | |
| 53442190 | |
| [Li+].[Li+].[Li+].CC(C)(COP(=O)([O-])OP(=O)([O-])OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)[O-])C(C(=O)NCCC(=O)NCCS)O |
| C21H33Li3N7O16P3S | |
| MFCD00075848 | |
| coenzyme a, trilithium salt | |
| trilithium;[5-(6-aminopurin-9-yl)-4-hydroxy-2-[[[[3-hydroxy-2,2-dimethyl-4-oxo-4-[[3-oxo-3-(2-sulfanylethylamino)propyl]amino]butoxy]-oxidophosphoryl]oxy-oxidophosphoryl]oxymethyl]oxolan-3-yl] hydrogen phosphate |
Safety and Handling
Recommended Storage : Store at -20 °C.