missing translation for 'onlineSavingsMsg'
Learn More

Coelenterazine, native, Aequorea sp., Thermo Scientific Chemicals

Catalog No. p-7076701
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66823MCR 1 mg
AAJ66823MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ66823MCR Supplier Thermo Scientific Chemicals Supplier No. J66823MCR
Only null left

A sensitive and specific chemiluminescence probe of the superoxide anion

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 55779-48-1
Molecular Formula C26H21N3O3
Molecular Weight (g/mol) 423.472
MDL Number MFCD00467176
InChI Key YHIPILPTUVMWQT-UHFFFAOYSA-N
Synonym coelenterazine, oplophorus luciferin, unii-3o1cb88rrd, clzn, coelenterate luciferin, coelenterazine, native, 3o1cb88rrd, 8-benzyl-2-4-hydroxybenzyl-6-4-hydroxyphenyl imidazo 1,2-a pyrazin-3 7h-one, 2-p-hydroxybenzyl-6-p-hydroxyphenyl-8-benzylimidazo 1,2-a pyrazin-3-7h-one, 3,2-dihydro-2-p-hydroxybenzyl-6-p-hydroxyphenyl-8-benzylimidazolo 1,2-a pyrazin-3-one
PubChem CID 2830
ChEBI CHEBI:2311
IUPAC Name 8-benzyl-6-(4-hydroxyphenyl)-2-[(4-hydroxyphenyl)methyl]-7H-imidazo[1,2-a]pyrazin-3-one
SMILES C1=CC=C(C=C1)CC2=C3N=C(C(=O)N3C=C(N2)C4=CC=C(C=C4)O)CC5=CC=C(C=C5)O

Specifications

Packaging Plastic bottle
Quantity 1 mg
Sensitivity Light and moisture sensitive
Solubility Information Soluble in methanol or ethanol
Formula Weight 423.46
Physical Form Solid
Chemical Name or Material Coelenterazine
TSCA No
Recommended Storage Store at -20°C; Store under Argon

RUO – Research Use Only