Learn More
Kent Scientific CODA™ Animal Holder
Allows researchers to constantly observe animal behavior during animal research.
$792.17
Specifications
| Construction | Adjustable black nose cone |
|---|---|
| For Use With (Application) | Observe Animal Behavior During Research |
| Type | Animal Holder |
| Catalog Number | Mfr. No. | For Use With (Equipment) | Size | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | For Use With (Equipment) | Size | Price | Quantity & Availability | ||||
13005156
|
Kent Scientific
HLDMS |
Mouse | Small |
Each for $792.17
|
|
||||
13005157
|
Kent Scientific
HLDMM |
Mouse | Medium |
Each for $792.17
|
|
||||
13005158
|
Kent Scientific
HLDML |
Mouse | X-Large |
Each for $792.17
|
|
||||
13005159
|
Kent Scientific
HLDRS |
Rat | Small |
Each for $792.17
|
|
||||
13005160
|
Kent Scientific
HLDRM |
Rat | Medium |
Each for $792.17
|
|
||||
13005161
|
Kent Scientific
HLDRL |
Rat | Large |
Each for $792.17
|
|
||||
13005162
|
Kent Scientific
HLDRX |
Rat | X-Large |
Each for $792.17
|
|
||||
Description
- Adjustable nose cone limits movement
- Helps animal remain comfortable
- Decreases injury to animal and researcher
- Dark black nose cone helps animal feel secure
- Nose protrudes through cone front, tail exits through rear hatch opening
Specifications
| Adjustable black nose cone | |
| Observe Animal Behavior During Research | |
| Animal Holder |