missing translation for 'onlineSavingsMsg'
Learn More

Cobaltocene, 98%

CAS: 1277-43-6 | C10H10Co | 189.12 g/mol

$132.78 - $132.78

Chemical Identifiers

CAS 1277-43-6
Molecular Formula C10H10Co
Molecular Weight (g/mol) 189.12
MDL Number MFCD00013749
InChI Key SNBRJOIYNQIWSZ-UHFFFAOYSA-N
Synonym Bis(cyclopentadienyl)cobalt
PubChem CID 24942376
IUPAC Name cobalt(2+);cyclopenta-1,3-diene
SMILES C1C=CC=[C-]1.C1C=CC=[C-]1.[Co+2]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC405320010
SDS View Documents
Thermo Scientific Chemicals
405320010
1 g Glass bottle
Each for $132.78
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1277-43-6
189.12
SNBRJOIYNQIWSZ-UHFFFAOYSA-N
24942376
C1C=CC=[C-]1.C1C=CC=[C-]1.[Co+2]
C10H10Co
MFCD00013749
Bis(cyclopentadienyl)cobalt
cobalt(2+);cyclopenta-1,3-diene

Specifications

1277-43-6
Black
Authentic
Glass bottle
MFCD00013749
Bis(cyclopentadienyl)cobalt
SNBRJOIYNQIWSZ-UHFFFAOYSA-N
cobalt(2+);cyclopenta-1,3-diene
24942376
98%
Cobaltocene
173°C
97.5% min.
98%
C10H10Co
1 g
Solubility in water: insoluble.,soluble in hydrocarbons
C1C=CC=[C-]1.C1C=CC=[C-]1.[Co+2]
189.12
189.12
Crystals
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable solid.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 215-061-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only