Learn More
Cobalt wire, 0.25mm (0.01in) dia, Puratronic™, 99.995% (metals basis)
CAS: 7440-48-4 | Co | 58.93 g/mol
$307.16 - $1085.66
Chemical Identifiers
| CAS | 7440-48-4 |
|---|---|
| Molecular Formula | Co |
| Molecular Weight (g/mol) | 58.93 |
| MDL Number | MFCD00010935 |
| InChI Key | GUTLYIVDDKVIGB-UHFFFAOYSA-N |
| Synonym | kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride |
| PubChem CID | 104730 |
| ChEBI | CHEBI:27638 |
| IUPAC Name | cobalt |
| SMILES | [Co] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7440-48-4 | |
| 58.93 | |
| GUTLYIVDDKVIGB-UHFFFAOYSA-N | |
| 104730 | |
| cobalt |
| Co | |
| MFCD00010935 | |
| kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride | |
| CHEBI:27638 | |
| [Co] |
Specifications
| 7440-48-4 | |
| Odorless | |
| Wire | |
| MFCD00010935 | |
| GUTLYIVDDKVIGB-UHFFFAOYSA-N | |
| cobalt | |
| 104730 | |
| 99.995% | |
| Cobalt wire, 0.25mm (0.01 in.) dia. |
| 1 m | |
| ≈0.44g/m | |
| Co | |
| kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride | |
| [Co] | |
| 58.93 | |
| CHEBI:27638 | |
| Puratronic | |
| (metals basis) |
Safety and Handling
GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
H317-H334-H350
EINECSNumber : 231-158-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only