missing translation for 'onlineSavingsMsg'
Learn More

Cobalt wire, 0.25mm (0.01in) dia, Puratronic™, 99.995% (metals basis)

Catalog No. AA10948G1
Change view
Click to view available options
Quantity:
1 m
5 m
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA10948G1 1 m
AA10948G5 5 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA10948G1 Supplier Thermo Scientific Chemicals Supplier No. 010948G1

CAS: 7440-48-4 | Co | 58.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-48-4
Molecular Formula Co
Molecular Weight (g/mol) 58.93
MDL Number MFCD00010935
InChI Key GUTLYIVDDKVIGB-UHFFFAOYSA-N
Synonym kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
PubChem CID 104730
ChEBI CHEBI:27638
IUPAC Name cobalt
SMILES [Co]

Specifications

Quantity 1 m
Odor Odorless
Weight ≈0.44g/m
Form Wire
Percent Purity 99.995%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Cobalt wire, 0.25mm (0.01 in.) dia.
Hazard Category H317-H334-H350
Hazard Statement GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
EINECSNumber 231-158-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only