missing translation for 'onlineSavingsMsg'
Learn More

Cobalt powder, -60 mesh, 99.5% (metals basis)

Catalog No. AA1045622
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1045622 100 g
AA1045636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1045622 Supplier Thermo Scientific Chemicals Supplier No. 01045622

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-48-4 | Co | 58.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-48-4
Molecular Formula Co
Molecular Weight (g/mol) 58.93
MDL Number MFCD00010935
InChI Key GUTLYIVDDKVIGB-UHFFFAOYSA-N
Synonym kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
PubChem CID 104730
ChEBI CHEBI:27638
IUPAC Name cobalt
SMILES [Co]

Specifications

Color Black
Appearance Black powder
Quantity 100 g
Mesh Size -60 Mesh
Odor Odorless
Form Powder
Assay Percent Range 99.5% (Metals basis)
Chemical Name or Material Cobalt
Hazard Category H228-H302-H317-H319-H330-H334-H335-H336-H350i-H360-H373-H500
Precautionary Statement P201-P202-P210-P240-P241-P260-P264b-P270-P271-P272-P280-P281-P284-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P333+P313-P363-P370+P378q-P501c
DOTInformation DOT Class: 4.1; Packaging Group: II
EINECSNumber 231-158-0
RTECSNumber GF8750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only