missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Cobalt powder, -400 mesh, 99.5% (metals basis)

Catalog No. AA4408518
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4408518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4408518 Supplier Thermo Scientific™ Supplier No. 4408518

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 7440-48-4
Quantity 50 g
Form Solid
MDL Number MFCD00010935
UN Number UN3089
Percent Purity 99.5%