Learn More
Cobalt powder, -22 mesh, Puratronic™, 99.998% (metals basis)
CAS: 7440-48-4 | Co | 58.93 g/mol
$204.78 - $713.18
Chemical Identifiers
| CAS | 7440-48-4 |
|---|---|
| Molecular Formula | Co |
| Molecular Weight (g/mol) | 58.93 |
| MDL Number | MFCD00010935 |
| InChI Key | GUTLYIVDDKVIGB-UHFFFAOYSA-N |
| Synonym | kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride |
| PubChem CID | 104730 |
| ChEBI | CHEBI:27638 |
| IUPAC Name | cobalt |
| SMILES | [Co] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7440-48-4 | |
| 58.93 | |
| GUTLYIVDDKVIGB-UHFFFAOYSA-N | |
| 104730 | |
| cobalt |
| Co | |
| MFCD00010935 | |
| kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride | |
| CHEBI:27638 | |
| [Co] |
Specifications
| 7440-48-4 | |
| -22 Mesh | |
| Co | |
| kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride | |
| [Co] | |
| 58.93 | |
| CHEBI:27638 |
| 5 g | |
| 99.99% (metals basis) | |
| MFCD00010935 | |
| GUTLYIVDDKVIGB-UHFFFAOYSA-N | |
| cobalt | |
| 104730 | |
| Cobalt powder |
Safety and Handling
P201-P202-P210-P240-P241-P260-P264b-P270-P271-P272-P280-P281-P284-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P333+P313-P363-P370+P378q-P501c
H228-H302-H317-H319-H330-H334-H335-H336-H350i-H360-H373-H500
DOTInformation : Hazard Class: 4.1; Packaging Group: II
EINECSNumber : 231-158-0
RTECSNumber : GF8750000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only