missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt(IV) sulfide, 99.5% (metals basis), Thermo Scientific Chemicals
Description
Cobalt(IV) sulfide is a promising battery electrode material exhibiting good electrochemical properties for lithium sulfur batteries, thermal batteries and lithium ion secondary battery.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Black |
| Physical Form | Powder |
| Quantity | 5 g |
| UN Number | UN3077 |
| Solubility Information | Slightly soluble in acid. Insoluble in water. |
| IUPAC Name | cobalt(3+);trisulfide |
| Formula Weight | 123.06 |
| Percent Purity | 99.5% |
| Odor | Odorless |
| Assay | (metals basis excluding Ni), Ni <0.2% |
| Show More |
Safety and Handling
| Hazard Category | H317-H350 |
| Hazard Statement | GHS H Statement H317 May cause an allergic skin reaction. |
| Precautionary Statement | P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
| RTECSNumber | GG3310000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only