missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(III) fluoride, 99%

Catalog No. p-4407410
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1149014 25 g
AA1149022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1149014 Supplier Thermo Scientific Chemicals Supplier No. 01149014
Only null left

CAS: 10026-18-3 | CoF3

Cobalt(III) fluoride is a powerful fluorinating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10026-18-3
Molecular Formula CoF3
MDL Number MFCD00010942

Specifications

Color Brown
Physical Form Powder
Quantity 25 g
Assay Percent Range 99%
Solubility Information Insoluble in water.
Formula Weight 115.93
Odor Acrid
Sensitivity Moisture sensitive
Appearance Light brown powder
Density 3.88
Chemical Name or Material Cobalt(III) fluoride
Show More Show Less
Hazard Category H272-H302+H312+H332-H314
Precautionary Statement P210-P220-P221-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation DOT Class: 5.1, Packaging Group: III
EINECSNumber 233-062-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only