missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) tungsten oxide, 99%

pigment, drier for enamels, inks, and paints, electronic devices | CAS: 10101-58-3 | CoO4W | 306.77 g/mol

$92.85 - $926.78

Chemical Identifiers

CAS 10101-58-3
Molecular Formula CoO4W
Molecular Weight (g/mol) 306.77
MDL Number MFCD00016034
InChI Key BGLJOHZOBUGBBY-UHFFFAOYSA-N
Synonym cobalt ii tungsten oxide, cobalt ii tungstate, co.wo4, cobalt 2+ ;dioxido dioxo tungsten
PubChem CID 11738337
IUPAC Name cobalt(2+);dioxido(dioxo)tungsten
SMILES [O--].[O--].[O--].[O--].[Co++].[W]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4022309
View Documents
Thermo Scientific Chemicals
04022309
10 g
Each for $92.85
Only null left
 
AA4022318
View Documents
Thermo Scientific Chemicals
04022318
50 g
Each for $229.49
Only null left
 
AA4022330
View Documents
Thermo Scientific Chemicals
04022330
250 g
Each for $926.78
Only null left
 
Description

Description

It is used as a pigment, drier for enamels, inks, and paints, electronic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10101-58-3
306.77
BGLJOHZOBUGBBY-UHFFFAOYSA-N
11738337
[O--].[O--].[O--].[O--].[Co++].[W]
CoO4W
MFCD00016034
cobalt ii tungsten oxide, cobalt ii tungstate, co.wo4, cobalt 2+ ;dioxido dioxo tungsten
cobalt(2+);dioxido(dioxo)tungsten

Specifications

10101-58-3
10 g
MFCD00016034
Insoluble in water; Soluble in hot concentrated acids
[O--].[O--].[O--].[O--].[Co++].[W]
306.77
306.78
8.42 g/mL
Powder
CoO4W
cobalt ii tungsten oxide, cobalt ii tungstate, co.wo4, cobalt 2+ ;dioxido dioxo tungsten
BGLJOHZOBUGBBY-UHFFFAOYSA-N
cobalt(2+);dioxido(dioxo)tungsten
11738337
99%
Cobalt(II) tungsten oxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.

P201-P202-P281-P308+P313-P501c

H350

EINECSNumber : 233-254-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only