missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) tungsten oxide, 99%

Catalog No. AA4022309
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4022309 10 g
AA4022318 50 g
AA4022330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4022309 Supplier Thermo Scientific Chemicals Supplier No. 04022309

pigment, drier for enamels, inks, and paints, electronic devices | CAS: 10101-58-3 | CoO4W | 306.77 g/mol

It is used as a pigment, drier for enamels, inks, and paints, electronic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10101-58-3
Molecular Formula CoO4W
Molecular Weight (g/mol) 306.77
MDL Number MFCD00016034
InChI Key BGLJOHZOBUGBBY-UHFFFAOYSA-N
Synonym cobalt ii tungsten oxide, cobalt ii tungstate, co.wo4, cobalt 2+ ;dioxido dioxo tungsten
PubChem CID 11738337
IUPAC Name cobalt(2+);dioxido(dioxo)tungsten
SMILES [O--].[O--].[O--].[O--].[Co++].[W]

Specifications

Physical Form Powder
Quantity 10 g
Solubility Information Insoluble in water; Soluble in hot concentrated acids
Formula Weight 306.78
Percent Purity 99%
Density 8.42 g/mL
Chemical Name or Material Cobalt(II) tungsten oxide
Hazard Category H350
Hazard Statement GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 233-254-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only