Learn More
Cobalt(II) thiocyanate, 98+%
CAS: 3017-60-5 | C2H2CoN2S2 | 177.105 g/mol
Supplier: Thermo Scientific Chemicals B2346030
Description
Cobalt(II) thiocyanate is best known for Scott test, which is a proven screening test for the presence of cocaine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCobalt(II) thiocyanate is best known for Scott test, which is a proven screening test for the presence of cocaine.
Solubility
Soluble in water.
Notes
Incompatible with acids and strong oxidizing agents.
Specifications
| Cobalt(II) thiocyanate | |
| Odorless | |
| Co(SCN)2 | |
| 250 g | |
| Soluble in water. | |
| C(#N)S.C(#N)S.[Co] | |
| 177.105 | |
| 175.1 |
| 3017-60-5 | |
| C2H2CoN2S2 | |
| MFCD00010937 | |
| UN3077 | |
| IQHZUTZWNWRODJ-UHFFFAOYSA-N | |
| cobalt;thiocyanic acid | |
| 131877393 | |
| ≥98% |
Chemical Identifiers
| 3017-60-5 | |
| 177.105 | |
| IQHZUTZWNWRODJ-UHFFFAOYSA-N | |
| cobalt;thiocyanic acid |
| C2H2CoN2S2 | |
| MFCD00010937 | |
| 131877393 | |
| C(#N)S.C(#N)S.[Co] |
Safety and Handling
GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
H302+H312+H332-H350
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 221-156-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only