missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) thiocyanate, 98+%

CAS: 3017-60-5 | C2H2CoN2S2 | 177.105 g/mol

Supplier:  Thermo Scientific Chemicals B2346018

 View more versions of this product

Catalog No. AAB2346018

Only null left
Explore available promotions

Description

Description

Cobalt(II) thiocyanate is best known for Scott test, which is a proven screening test for the presence of cocaine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cobalt(II) thiocyanate is best known for Scott test, which is a proven screening test for the presence of cocaine.

Solubility
Soluble in water.

Notes
Incompatible with acids and strong oxidizing agents.
Specifications

Specifications

Cobalt(II) thiocyanate
Odorless
Co(SCN)2
50 g
Soluble in water.
C(#N)S.C(#N)S.[Co]
177.105
175.1
3017-60-5
C2H2CoN2S2
MFCD00010937
UN3077
IQHZUTZWNWRODJ-UHFFFAOYSA-N
cobalt;thiocyanic acid
131877393
≥98%

Chemical Identifiers

3017-60-5
177.105
IQHZUTZWNWRODJ-UHFFFAOYSA-N
cobalt;thiocyanic acid
C2H2CoN2S2
MFCD00010937
131877393
C(#N)S.C(#N)S.[Co]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c

H302+H312+H332-H350

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 221-156-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only