Learn More
Cobalt(II) phthalocyanine
CAS: 3317-67-7 | C32H16CoN8 | 571.469 g/mol
Supplier: Thermo Scientific Chemicals 04149606
Description
Cobalt(II) phthalocyanine may be used to develop carbon black based electrocatalysts. CoPC may be used as an end capping agent of a hyperbranched poly(aryl ether ketone) to be used for oxidative decomposition of 2,4,6,-trichlorophenol. CoPC modified carbon paste may be used as indicators for electrocatalytic amperometric measurements. ZnO impregnated with CoPC may be used as a sensitizer to determine the photodegradation of cyanide in aqueous suspension. CoPC catalyzed aerobic regenerations of aldehydes and ketones have been investigated.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Cobalt(II) phthalocyanine | |
| Blue to Purple | |
| C32H16CoN8 | |
| 5 g | |
| Insoluble in water. | |
| C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.[Co+2] | |
| 2734991 | |
| Powder |
| 3317-67-7 | |
| Odorless | |
| MFCD00010718 | |
| cobalt ii phthalocyanine, cobalt phthalocyanine, phthalocyanine cobalt ii, phthalocyanine, cobalt, cobalt ii phthalocyanine, beta-form, dye content 97 % | |
| MPMSMUBQXQALQI-UHFFFAOYSA-N | |
| 571.469 | |
| 571.47 |
Chemical Identifiers
| 3317-67-7 | |
| 571.469 | |
| MPMSMUBQXQALQI-UHFFFAOYSA-N | |
| 2734991 |
| C32H16CoN8 | |
| MFCD00010718 | |
| cobalt ii phthalocyanine, cobalt phthalocyanine, phthalocyanine cobalt ii, phthalocyanine, cobalt, cobalt ii phthalocyanine, beta-form, dye content 97 % | |
| C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.[Co+2] |
Safety and Handling
GHS H Statement
H341-H317
Suspected of causing genetic defects.
May cause an allergic skin reaction.
P201-P202-P261-P281-P285-P304+P341-P308+P313-P342+P311-P501c
H334-H341-H350
EINECSNumber : 222-012-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only