missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) phthalocyanine

Catalog No. AA4149614
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4149614 25 g
AA4149603 1 g
AA4149606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4149614 Supplier Thermo Scientific Chemicals Supplier No. 04149614

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3317-67-7 | C32H16CoN8 | 571.469 g/mol

Cobalt(II) phthalocyanine may be used to develop carbon black based electrocatalysts. CoPC may be used as an end capping agent of a hyperbranched poly(aryl ether ketone) to be used for oxidative decomposition of 2,4,6,-trichlorophenol. CoPC modified carbon paste may be used as indicators for electrocatalytic amperometric measurements. ZnO impregnated with CoPC may be used as a sensitizer to determine the photodegradation of cyanide in aqueous suspension. CoPC catalyzed aerobic regenerations of aldehydes and ketones have been investigated.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3317-67-7
Molecular Formula C32H16CoN8
Molecular Weight (g/mol) 571.469
MDL Number MFCD00010718
InChI Key MPMSMUBQXQALQI-UHFFFAOYSA-N
Synonym cobalt ii phthalocyanine, cobalt phthalocyanine, phthalocyanine cobalt ii, phthalocyanine, cobalt, cobalt ii phthalocyanine, beta-form, dye content 97 %
PubChem CID 2734991
SMILES C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.[Co+2]

Specifications

Color Blue to Purple
Odor Odorless
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 571.47
Physical Form Powder
Chemical Name or Material Cobalt(II) phthalocyanine
Hazard Category H334-H341-H350
Hazard Statement GHS H Statement
H341-H317
Suspected of causing genetic defects.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P281-P285-P304+P341-P308+P313-P342+P311-P501c
EINECSNumber 222-012-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only