missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) phosphate octahydrate

Catalog No. p-7049815
Change view
Click to view available options
Quantity:
250 g
50 g
Catalog No. Quantity
AA2311118 50 g
AA2311130 250 g
2 options

Catalog No. AA2311118

Supplier: Thermo Scientific Chemicals 02311118

Only null left

CAS: 10294-50-5 | Co3H16O16P2 | 510.859 g/mol

It is used as a pigment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10294-50-5
Molecular Formula Co3H16O16P2
Molecular Weight (g/mol) 510.859
MDL Number MFCD00016028
InChI Key QOQSIXJUYVUEMP-UHFFFAOYSA-H
Synonym cobalt ii phosphate octahydrate, cobaltous phosphate octahydrate, cobalt phosphate octahydrate, phosphoric acid, cobalt 2+ salt 2:3 , octahydrate, acmc-20ale3, 3co.2po4.8h2o, cobalt 2+ diphosphate octahydrate
PubChem CID 202398
IUPAC Name cobalt(2+);diphosphate;octahydrate
SMILES O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Co+2].[Co+2].[Co+2]

Specifications

Physical Form Powder
Quantity 50 g
Solubility Information Slightly soluble in water. Soluble in mineral acids.
Formula Weight 510.87 (366.74 Anhydrous)
Odor Odorless
Density 2.769 g/mL
Chemical Name or Material Cobalt(II) phosphate octahydrate
Hazard Category H302-H315-H317-H319-H334-H335-H350
Hazard Statement GHS H Statement
H301-H341-H317
Toxic if swallowed.
Suspected of causing genetic defects.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P272-P280-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P342+P311-P362-P501c
EINECSNumber 236-655-6
RTECSNumber TB9279500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only