missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) perchlorate hexahydrate, 99.999% (metals basis)

Catalog No. p-7059270
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4432009 10 g
AA4432018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4432009 Supplier Thermo Scientific Chemicals Supplier No. 04432009
Only null left

CAS: 13478-33-6 | Cl2CoO8 | 257.825 g/mol

It is used as a chemical and organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13478-33-6
Molecular Formula Cl2CoO8
Molecular Weight (g/mol) 257.825
MDL Number MFCD00150207
InChI Key BSUSEPIPTZNHMN-UHFFFAOYSA-L
Synonym cobaltous perchlorate, cobalt diperchlorate, cobalt ii perchlorate, cobalt diperchlorate hexahydrate, cobalt perchlorate hexahydrate, cobaltous perchlorate, hexahydrate, cobalt ii perchlorate, hexahydrate, perchloric acid, cobalt ii salt, hexahydrate, diperchloratocobalt
PubChem CID 26064
IUPAC Name cobalt(2+);diperchlorate
SMILES [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Co+2]

Specifications

Physical Form Crystalline
Quantity 10 g
UN Number UN1481
Solubility Information Soluble in water.
Formula Weight 366.74 (257.83 Anhydrous)
Percent Purity 99.999%
Odor Odorless
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Cobalt (II) perchlorate hexahydrate
Hazard Category H272-H315-H319-H334-H335-H350
Hazard Statement GHS H Statement
H272-H351-H315-H319-H317-H335
May intensify fire
oxidizer.
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P220-P221-P261-P264b-P271-P280i-P281-P285-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P342+P311-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
EINECSNumber 236-653-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only