missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) oxide, 99.995% (metals basis)

Catalog No. p-7059297
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4435406 5 g
AA4435414 25 g
AA4435422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4435406 Supplier Thermo Scientific Chemicals Supplier No. 04435406
Only null left

CAS: 1307-96-6 | CoO | 74.93 g/mol

Cobalt(II) oxide is widely used in the ceramic industry. It is an additive to create blue-colored glazes and enamels. It finds application in the manufacture of magnetic and electronic component materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1307-96-6
Molecular Formula CoO
Molecular Weight (g/mol) 74.93
MDL Number MFCD00016031
InChI Key IVMYJDGYRUAWML-UHFFFAOYSA-N
Synonym cobalt ii oxide, zaffre, cobalt monoxide, cobalt black, cobalt oxide, cobaltous oxide, cobalt monooxide, monocobalt oxide, cobalt 2+ oxide, ci pigment black 13
PubChem CID 14786
IUPAC Name oxocobalt
SMILES O=[Co]

Specifications

Melting Point 1935°C
Color Brown-Green
Physical Form Powder
Quantity 5 g
UN Number UN2811
Merck Index 14,2446
Solubility Information Insoluble in water.
Formula Weight 74.93
Percent Purity 99.995%
Concentration or Composition (by Analyte or Components) CoO, may contain up to 10% Co3O4
Sensitivity Air Sensitive
Density 6.45 g/mL
Assay (metals basis)
Chemical Name or Material Cobalt (II) oxide
Show More Show Less
Hazard Category H301-H317-H330-H334-H350
Hazard Statement GHS H Statement
H351-H302-H317
Suspected of causing cancer.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P284-P285-P301+P310-P302+P352-P304+P340-P308+P313-P310-P330-P333+P313-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 215-154-6
RTECSNumber GG2800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only