missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt(Ii) Nitrate Hexahydrate, Honeywell Fluka™
ACS Reagent,≥98%
Supplier: Honeywell Chemicals 239267100G
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Cobalt(Ii) Nitrate Hexahydrate | |
| 100 g | |
| Co(NO3)2 · 6H2O | |
| 1477 | |
| QGUAJWGNOXCYJF-UHFFFAOYSA-N | |
| cobalt(2+);dinitrate;hexahydrate | |
| 24821 | |
| 291.03g/mol | |
| ACS Reagent |
| 10026-22-9 | |
| CoH12N2O12 | |
| MFCD00149647 | |
| cobalt nitrate hexahydrate, cobalt ii nitrate hexahydrate, cobaltous nitrate hexahydrate, cobalt dinitrate hexahydrate, cobalt 2+ nitrate hexahydrate, cobaltous nitrate, hexahydrate, kobalt ii-nitrat-hexahydrat, nitric acid, cobalt 2+ salt, hexahydrate | |
| O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
| 291.03 | |
| CHEBI:86214 | |
| ≥98% | |
| Plastic Bottle |
Chemical Identifiers
| 10026-22-9 | |
| 291.03 | |
| QGUAJWGNOXCYJF-UHFFFAOYSA-N | |
| 24821 | |
| cobalt(2+);dinitrate;hexahydrate |
| CoH12N2O12 | |
| MFCD00149647 | |
| cobalt nitrate hexahydrate, cobalt ii nitrate hexahydrate, cobaltous nitrate hexahydrate, cobalt dinitrate hexahydrate, cobalt 2+ nitrate hexahydrate, cobaltous nitrate, hexahydrate, kobalt ii-nitrat-hexahydrat, nitric acid, cobalt 2+ salt, hexahydrate | |
| CHEBI:86214 | |
| O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Safety and Handling
P201-P210-P261-P280-P284-P304 + P340-P308 + P313
H272-H302-H317-H334-H341-H350i-H360F-H410
RTECSNumber : QU7355500