missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) nitrate hexahydrate, ACS, 98.0-102.0%

Catalog No. AA3641822
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3641822 100 g
AA3641836 500 g
AA36418A4 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3641822 Supplier Thermo Scientific Chemicals Supplier No. 03641822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Use As a Battery Additive, in the Manufacture of Cobalt Pigments and Invisible Inks, For Decorating Stoneware and Porcelain | CAS: 10026-22-9 | CoH12N2O12 | 291.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications:
  • As battery additive, in manufacture of cobalt pigments and invisible inks, for decorating stoneware and procelain, and in preparation of catalysts

Chemical Identifiers

CAS 10026-22-9
Molecular Formula CoH12N2O12
Molecular Weight (g/mol) 291.03
MDL Number MFCD00149647
InChI Key QGUAJWGNOXCYJF-UHFFFAOYSA-N
Synonym cobalt nitrate hexahydrate, cobalt ii nitrate hexahydrate, cobaltous nitrate hexahydrate, cobalt dinitrate hexahydrate, cobalt 2+ nitrate hexahydrate, cobaltous nitrate, hexahydrate, kobalt ii-nitrat-hexahydrat, nitric acid, cobalt 2+ salt, hexahydrate
PubChem CID 24821
ChEBI CHEBI:86214
SMILES O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 55°C to 56°C
Color Red
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 98.0-102.0%
Solubility Information Very soluble in water,alcohol,and most organic solvents.
IUPAC Name λ²-cobalt(2+) hexahydrate dinitrate
Formula Weight 291.04 (182.94 Anhydrous)
Grade ACS
Odor Odorless
Refractive Index 1.52
Appearance Red crystalline powder
Density 1.87 g/cm3
Chemical Name or Material Cobalt(II) nitrate hexahydrate, For analysis ACS
Show More Show Less
Hazard Category H272-H302+H332-H317-H318-H334-H341-H350i-H360F
Precautionary Statement P201-P202-P210-P220-P221-P261-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P333+P313-P342+P311-P363-P370+P378q-P501c
DOTInformation DOT Class: 5.1, Packaging Group: III
EINECSNumber 233-402-1
RTECSNumber QU7355500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only