Learn More
Cobalt(II) nitrate hexahydrate, 98+%, ACS reagent
CAS: 10026-22-9 | CoH12N2O12 | 291.03 g/mol
Supplier: Thermo Scientific Chemicals 423581000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Cobalt(II) nitrate hexahydrate | |
| 10026-22-9 | |
| Adhering Crystalline Powder or Crystals | |
| 98.0 to 102% (Complexometry) | |
| Co(NO3)2·6H2O | |
| 15, 2428 | |
| Solubility in water: 1340g/L. Other solubilities: soluble in ethanol and acetone, soluble in most common organic solvents | |
| O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
| 291.03 | |
| CHEBI:86214 | |
| ≥98% | |
| Plastic bottle | |
| 1.8800g/mL |
| 55.0°C | |
| Brown-Pink to Red | |
| 100 g | |
| CoH12N2O12 | |
| MFCD00149647 | |
| cobalt nitrate hexahydrate, cobalt ii nitrate hexahydrate, cobaltous nitrate hexahydrate, cobalt dinitrate hexahydrate, cobalt 2+ nitrate hexahydrate, cobaltous nitrate, hexahydrate, kobalt ii-nitrat-hexahydrat, nitric acid, cobalt 2+ salt, hexahydrate | |
| QGUAJWGNOXCYJF-UHFFFAOYSA-N | |
| cobalt(2+);dinitrate;hexahydrate | |
| 24821 | |
| 291.02 | |
| ACS Reagent | |
| 1.88 |
Chemical Identifiers
| 10026-22-9 | |
| 291.03 | |
| QGUAJWGNOXCYJF-UHFFFAOYSA-N | |
| 24821 | |
| cobalt(2+);dinitrate;hexahydrate |
| CoH12N2O12 | |
| MFCD00149647 | |
| cobalt nitrate hexahydrate, cobalt ii nitrate hexahydrate, cobaltous nitrate hexahydrate, cobalt dinitrate hexahydrate, cobalt 2+ nitrate hexahydrate, cobaltous nitrate, hexahydrate, kobalt ii-nitrat-hexahydrat, nitric acid, cobalt 2+ salt, hexahydrate | |
| CHEBI:86214 | |
| O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Safety and Handling
GHS H Statement
May intensify fire; oxidizer.
Harmful if swallowed.
Harmful if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Causes serious ey
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remov
GHS Signal Word: Danger
RTECSNumber : QU7355500
RUO – Research Use Only