missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) nitrate hexahydrate, 98+%, ACS reagent

Catalog No. AC423580050
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423580050 5 g Glass Bottle
AC423581000 100 g Plastic bottle
AC423585000 500 g Plastic bottle
AC423580025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC423580050 Supplier Thermo Scientific Chemicals Supplier No. 423580050
Only null left

CAS: 10026-22-9 | CoH12N2O12 | 291.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10026-22-9
Molecular Formula CoH12N2O12
Molecular Weight (g/mol) 291.03
MDL Number MFCD00149647
InChI Key QGUAJWGNOXCYJF-UHFFFAOYSA-N
Synonym cobalt nitrate hexahydrate, cobalt ii nitrate hexahydrate, cobaltous nitrate hexahydrate, cobalt dinitrate hexahydrate, cobalt 2+ nitrate hexahydrate, cobaltous nitrate, hexahydrate, kobalt ii-nitrat-hexahydrat, nitric acid, cobalt 2+ salt, hexahydrate
PubChem CID 24821
ChEBI CHEBI:86214
IUPAC Name cobalt(2+);dinitrate;hexahydrate
SMILES O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 55.0°C
Color Brown-Pink to Red
Physical Form Adhering Crystalline Powder or Crystals
Quantity 5 g
Assay Percent Range 98.0 to 102% (Complexometry)
Linear Formula Co(NO3)2·6H2O
Merck Index 15, 2428
Solubility Information Solubility in water: 1340g/L. Other solubilities: soluble in ethanol and acetone, soluble in most common organic solvents
Formula Weight 291.02
Percent Purity ≥98%
Grade ACS Reagent
Packaging Glass Bottle
Specific Gravity 1.88
Density 1.8800g/mL
Chemical Name or Material Cobalt(II) nitrate hexahydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May intensify fire; oxidizer.
Harmful if swallowed.
Harmful if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Causes serious ey
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remov
RTECSNumber QU7355500

RUO – Research Use Only