missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) nitrate hexahydrate, 97.7% min

CAS: 10026-22-9 | CoH12N2O12 | 291.03 g/mol

Supplier:  Thermo Scientific Chemicals 011341A1

 View more versions of this product

Catalog No. AA11341A1

Only null left
Explore available promotions

Description

Description

Cobalt(II) nitrate hexahydrate is used in the preparation of metallic cobalt and cobalt based dyes and pigments. It is a precursor for the preparation of catalyst for use in certain organic reactions. It finds use in catalysis, vitamin preparations, and porcelain decoration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cobalt(II) nitrate hexahydrate
10026-22-9
Crystalline
CoH12N2O12
UN1477
cobalt nitrate hexahydrate, cobalt ii nitrate hexahydrate, cobaltous nitrate hexahydrate, cobalt dinitrate hexahydrate, cobalt 2+ nitrate hexahydrate, cobaltous nitrate, hexahydrate, kobalt ii-nitrat-hexahydrat, nitric acid, cobalt 2+ salt, hexahydrate
QGUAJWGNOXCYJF-UHFFFAOYSA-N
cobalt(2+);dinitrate;hexahydrate
24821
291.04 (182.94 Anhydrous)
Odorless
1.87 g/mL
55°C to 56°C
Red
1 kg
MFCD00149647
14,2444
Soluble in water,alcohol,acetone and ammonia.
O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
291.03
CHEBI:86214
≤97.7%
1.52

Chemical Identifiers

10026-22-9
291.03
QGUAJWGNOXCYJF-UHFFFAOYSA-N
24821
cobalt(2+);dinitrate;hexahydrate
CoH12N2O12
MFCD00149647
cobalt nitrate hexahydrate, cobalt ii nitrate hexahydrate, cobaltous nitrate hexahydrate, cobalt dinitrate hexahydrate, cobalt 2+ nitrate hexahydrate, cobaltous nitrate, hexahydrate, kobalt ii-nitrat-hexahydrat, nitric acid, cobalt 2+ salt, hexahydrate
CHEBI:86214
O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H334-H350-H360-H341-H302-H317
May intensify fire; oxidizer.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
May cause an allergic skin reaction.

P201-P202-P210-P220-P221-P261-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P333+P313-P342+P311-P363-P370+P378q-P501c

H272-H302+H332-H317-H318-H334-H341-H350i-H360F

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.

EINECSNumber : 233-402-1

RTECSNumber : QU7355500

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only