missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II,III) oxide

Catalog No. AAA1612130
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1612130 250 g
AAA161210B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1612130 Supplier Thermo Scientific Chemicals Supplier No. A1612130

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1308-06-1 | Co3O4 | 240.80 g/mol

Cobalt(II,III) oxide is currently utilized in the process of artificial photosynthesis. It is also used for glass, optic and ceramic applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cobalt(II,III) oxide is currently utilized in the process of artificial photosynthesis. It is also used for glass, optic and ceramic applications.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1308-06-1
Molecular Formula Co3O4
Molecular Weight (g/mol) 240.80
MDL Number MFCD00010939
InChI Key UBEWDCMIDFGDOO-UHFFFAOYSA-N
Synonym oxocobalt; oxo oxocobaltiooxy cobalt, cobalt ii oxide; cobalt iii oxide, cobalt ii, coo.co2o3, aronis24129, cobaltic tetraoxide,, cobalt ii,iii oxide, powder, <10 mum, cobalt ii oxide; oxo oxocobaltio oxy cobalt, cobalt ii,iii oxide metals basis , 2-6 micron powder, oxidanylidenecobalt; oxidanylidene oxidanylidenecobaltiooxy cobalt
PubChem CID 11651651
IUPAC Name oxocobalt;oxo(oxocobaltiooxy)cobalt
SMILES [O--].[O--].[O--].[O--].[Co++].[Co+3].[Co+3]

Specifications

Melting Point 900°C (decomposition)
Quantity 250 g
Density 6.11
UN Number UN3077
Merck Index 14,2429
Solubility Information Insoluble in water.
Formula Weight 240.8
Chemical Name or Material Cobalt(II,III) oxide
Hazard Category H334-H350i
Hazard Statement GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P281-P285-P304+P341-P308+P313-P342+P311-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 215-157-2
RTECSNumber GG2500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only