missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II,III) oxide, 99.7% (metals basis)

Catalog No. AA4018414
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4018414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4018414 Supplier Thermo Scientific Chemicals Supplier No. 04018414

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Use With Enamels, Semiconductors, Grinding Wheels | CAS: 1308-06-1 | Co3O4 | 240.80 g/mol

Finds use in enamels, semiconductors, grinding wheels. Used in catalysis, superconductors, ceramics and other fields as an important inorganic material. Also used as catalyst and catalyst carrier, as electrode active material, for glass, porcelain colorants and pigments, as chemical industry oxidant and as a catalyst for organic synthesis, in senior goggles and other filter materials, in temperature and gas sensors, in electronic ceramics, in lithium ion battery electrode materials, magnetic materials, electrochromic devices, enamels, solar energy absorbers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1308-06-1
Molecular Formula Co3O4
Molecular Weight (g/mol) 240.80
MDL Number MFCD00010939
InChI Key UBEWDCMIDFGDOO-UHFFFAOYSA-N
Synonym oxocobalt; oxo oxocobaltiooxy cobalt, cobalt ii oxide; cobalt iii oxide, cobalt ii, coo.co2o3, aronis24129, cobaltic tetraoxide,, cobalt ii,iii oxide, powder, <10 mum, cobalt ii oxide; oxo oxocobaltio oxy cobalt, cobalt ii,iii oxide metals basis , 2-6 micron powder, oxidanylidenecobalt; oxidanylidene oxidanylidenecobaltiooxy cobalt
PubChem CID 11651651
SMILES [O--].[O--].[O--].[O--].[Co++].[Co+3].[Co+3]

Specifications

Melting Point 900°C (decomposition)
Color Black
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.7% (Metals basis)
Solubility Information Soluble in acids and alkalis. Insoluble in water.
IUPAC Name oxocobalt;oxo(oxocobaltiooxy)cobalt
Formula Weight 240.8
Odor Odorless
Density 6.11
Chemical Name or Material Cobalt(II, III) oxide
Show More Show Less
Hazard Category H334-H350i
Hazard Statement GHS H Statement
H351-H317
Precautionary Statement P201-P202-P261-P281-P285-P304+P341-P308+P313-P342+P311-P501c
DOTInformation DOT Class: 9, Packaging Group: III
EINECSNumber 215-157-2
RTECSNumber GG2500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only