missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) gluconate hydrate

CAS: 1809162-99-9 | C12H26CoO15 | 469.258 g/mol

Supplier:  Thermo Scientific Chemicals B2429322

 View more versions of this product

Catalog No. AAB2429322


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cobalt(II) gluconate hydrate
1809162-99-9
C12H26CoO15
cobalt ii gluconate hydrate, 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanoyl oxy cobaltio 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanoate hydrate
C(C(C(C(C(C(=O)O)O)O)O)O)O.C(C(C(C(C(C(=O)O)O)O)O)O)O.O.[Co]
469.258
449.23 (Anhydrous)
>300°C
100 g
MFCD02684006
QBVYGKYWWCNZSO-XRDLMGPZSA-N
cobalt;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid;hydrate
117064710

Chemical Identifiers

1809162-99-9
469.258
QBVYGKYWWCNZSO-XRDLMGPZSA-N
117064710
C(C(C(C(C(C(=O)O)O)O)O)O)O.C(C(C(C(C(C(=O)O)O)O)O)O)O.O.[Co]
C12H26CoO15
MFCD02684006
cobalt ii gluconate hydrate, 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanoyl oxy cobaltio 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanoate hydrate
cobalt;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid;hydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H332-H317
Toxic if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.

P261-P264b-P270-P271-P272-P280g-P301+P312-P302+P352-P304+P340-P312-P330-P333+P313-P363-P501c

H302+H332-H317

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 276-206-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only