missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt(II) fluoride, 99%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| CAS | 10026-17-2 |
| Quantity | 25g |
| Assay Percent Range | 98.5% min. (ex F) |
| Molecular Formula | CoF2 |
| Linear Formula | CoF2 |
| MDL Number | MFCD00010941 |
| Merck Index | 15, 2424 |
| Synonym | cobalt ii fluoride, cobalt fluoride, cobalt difluoride, cobaltous fluoride, hsdb 236, cobalt fluoride pink, cobalt ii fluoride, powder, cobalt ii fluoride, anhydrous, cobalt ii fluoride, ultra dry |
| InChI Key | YCYBZKSMUPTWEE-UHFFFAOYSA-L |
| SMILES | F[Co]F |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement: Causes serious eye damage. Causes severe skin burns and eye damage. |
| Precautionary Statement | GHS P Statement: IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 233-061-9 |
| RTECSNumber | GG0770000 |
| TSCA | TSCA |
RUO – Research Use Only