missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) chloride, 97%, anhydrous

Catalog No. p-46430
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC214130050 5 g Glass Bottle
AC214131000 100 g Plastic bottle
AC214135000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC214130050 Supplier Thermo Scientific Chemicals Supplier No. 214130050
Only null left

CAS: 7646-79-9 | Cl2Co | 129.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 7646-79-9
Molecular Formula Cl2Co
Molecular Weight (g/mol) 129.83
MDL Number MFCD00010938
InChI Key GVPFVAHMJGGAJG-UHFFFAOYSA-L
Synonym cobalt ii chloride, cobaltchloride, cobalt chloride 0.1 m solution, cobalt ii chloride, anhydrous, cobalt ii-chloride, curator_000013, cobalt chloride anhydrous, wln: co g2, cobalt-ii-chloride
IUPAC Name λ²-cobalt(2+) dichloride
SMILES [Cl-].[Cl-].[Co++]

Specifications

Boiling Point 1049.0°C
Melting Point 735.0°C
Color Blue
Physical Form Powder
Quantity 5 g
Assay Percent Range 97%
Linear Formula CoCl2
Fieser 01,155; 06,127; 08,291; 10,101; 13,249; 14,99; 15,97
Merck Index 15, 2421
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and acetone
Formula Weight 129.84
Percent Purity 97%
Packaging Glass Bottle
Density 3.35
Chemical Name or Material Cobalt(II) chloride, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause cancer by inhalation.
Suspected of causing genetic defects.
May damage fertility.
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cau
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
IF
EINECSNumber 231-589-4
RTECSNumber GF9800000
TSCA TSCA

RUO – Research Use Only