Learn More
Cobalt(II) carbonate, 99% (metals basis), Co 45% min
For Use in Ceramics, Manufacturing Co Compounds, Pigments | CAS: 513-79-1 | CCoO3 | 118.94 g/mol
$159.04 - $390.14
Chemical Identifiers
| CAS | 513-79-1 |
|---|---|
| Molecular Formula | CCoO3 |
| Molecular Weight (g/mol) | 118.94 |
| MDL Number | MFCD00010940 |
| InChI Key | ZOTKGJBKKKVBJZ-UHFFFAOYSA-L |
| Synonym | cobalt carbonate, cobalt ii carbonate, cobaltous carbonate, sphaerocobaltite, cobalt spar, cobalt 2+ carbonate, cobalt monocarbonate, cobalt carbonate 1:1, cobalt carbonate coco3, unii-7h73a68fuv |
| PubChem CID | 10565 |
| IUPAC Name | cobalt(2+);carbonate |
| SMILES | [Co++].[O-]C([O-])=O |
Description
Cobalt(II) carbonate is precursor to cobalt carbonyl, cobalt salts and blue pottery glazes such as delftware. It is used as an essential element in dietary supplements and also used in the manufacture of paints and pigments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 513-79-1 | |
| 118.94 | |
| ZOTKGJBKKKVBJZ-UHFFFAOYSA-L | |
| 10565 | |
| [Co++].[O-]C([O-])=O |
| CCoO3 | |
| MFCD00010940 | |
| cobalt carbonate, cobalt ii carbonate, cobaltous carbonate, sphaerocobaltite, cobalt spar, cobalt 2+ carbonate, cobalt monocarbonate, cobalt carbonate 1:1, cobalt carbonate coco3, unii-7h73a68fuv | |
| cobalt(2+);carbonate |
Specifications
| (decomposition) | |
| Pink | |
| 100 g | |
| CCoO3 | |
| cobalt carbonate, cobalt ii carbonate, cobaltous carbonate, sphaerocobaltite, cobalt spar, cobalt 2+ carbonate, cobalt monocarbonate, cobalt carbonate 1:1, cobalt carbonate coco3, unii-7h73a68fuv | |
| ZOTKGJBKKKVBJZ-UHFFFAOYSA-L | |
| cobalt(2+);carbonate | |
| 10565 | |
| Odorless | |
| Cobalt(II) carbonate |
| 513-79-1 | |
| Powder | |
| 99% (Metals basis, Co 45% min) | |
| MFCD00010940 | |
| Insoluble in water,alcohol and methyl acetate. | |
| [Co++].[O-]C([O-])=O | |
| 118.94 | |
| 118.94 | |
| 4.13 |
Safety and Handling
P201-P202-P261-P264b-P270-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P341-P308+P313-P330-P333+P313-P342+P311-P363-P501c
H302-H317-H334-H341-H350i-H360F
DOTInformation : DOT Class: 9, Packaging Group: III
EINECSNumber : 208-169-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only