missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt(II) carbonate, 99% (metals basis), Co 45% min
Description
Cobalt(II) carbonate is precursor to cobalt carbonyl, cobalt salts and blue pottery glazes such as delftware. It is used as an essential element in dietary supplements and also used in the manufacture of paints and pigments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | (decomposition) |
| Color | Pink |
| Physical Form | Powder |
| Quantity | 100 g |
| Assay Percent Range | 99% (Metals basis, Co 45% min) |
| Solubility Information | Insoluble in water,alcohol and methyl acetate. |
| Formula Weight | 118.94 |
| Odor | Odorless |
| Density | 4.13 |
| Chemical Name or Material | Cobalt(II) carbonate |
Safety and Handling
| Hazard Category | H302-H317-H334-H341-H350i-H360F |
| Precautionary Statement | P201-P202-P261-P264b-P270-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P341-P308+P313-P330-P333+P313-P342+P311-P363-P501c |
| DOTInformation | DOT Class: 9, Packaging Group: III |
| EINECSNumber | 208-169-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only