missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) carbonate, 99.5% (metals basis)

CAS: 513-79-1 | CCoO3 | 118.94 g/mol

Supplier:  Thermo Scientific Chemicals 04274530

 View more versions of this product

Catalog No. AA4274530


Only null left
Explore available promotions

Description

Description

Cobalt(II) carbonate is a precursor to cobalt carbonyl, cobalt salts and blue pottery glazes such as delftware. It is used as an essential element in dietary supplements and is also used in the manufacture of paints and pigments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cobalt(II) carbonate
513-79-1
250 g
CCoO3
cobalt carbonate, cobalt ii carbonate, cobaltous carbonate, sphaerocobaltite, cobalt spar, cobalt 2+ carbonate, cobalt monocarbonate, cobalt carbonate 1:1, cobalt carbonate coco3, unii-7h73a68fuv
ZOTKGJBKKKVBJZ-UHFFFAOYSA-L
cobalt(2+);carbonate
10565
4.13 g/mL
(decomposition)
Powder
99.5% (metals basis)
MFCD00010940
Insoluble in water,alcohol and methyl acetate.
[Co++].[O-]C([O-])=O
118.94
118.94

Chemical Identifiers

513-79-1
118.94
ZOTKGJBKKKVBJZ-UHFFFAOYSA-L
10565
[Co++].[O-]C([O-])=O
CCoO3
MFCD00010940
cobalt carbonate, cobalt ii carbonate, cobaltous carbonate, sphaerocobaltite, cobalt spar, cobalt 2+ carbonate, cobalt monocarbonate, cobalt carbonate 1:1, cobalt carbonate coco3, unii-7h73a68fuv
cobalt(2+);carbonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P201-P202-P261-P264b-P270-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P341-P308+P313-P330-P333+P313-P342+P311-P363-P501c

H302-H317-H334-H341-H350i-H360F

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 208-169-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only