Learn More
Cobalt(II) bromide hydrate
Description
Hygrometers, catalystsCobalt(II) bromide hydrate is mainly used as a catalyst in organic synthesis, like C-metal, C-heteroatom and C-C bond formations, addition to unsaturated bonds, cyclizations, polymerization, oxidations reductions and carbonylations. It reacts with ammonia to form coordination compound bromopentaamminecobalt(III) bromide. It reacts with triphenylphosphine and form complexes of cobalt(II) bromide, which used as a catalyst in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 47°C to 48°C (decomposition) |
| Color | Purple-Red |
| Physical Form | Crystalline Granules |
| Quantity | 50 g |
| Solubility Information | Soluble in water. |
| IUPAC Name | λ²-cobalt(2+) dibromide |
| Formula Weight | 218.75 (Anhydrous) |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Appearance | Red-purple crystalline granules |
| Show More |
Safety and Handling
| Hazard Category | H302-H315-H319-H335 |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| DOTInformation | Hazard Class: 9; Packaging Group: III |
| EINECSNumber | 232-166-7 |
| RTECSNumber | GF9595000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only