Learn More
Cobalt(II) bromide hydrate, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 318541000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Cobalt(II) bromide hydrate | |
| 100g | |
| CoBr2·xH2O | |
| cobalt ii bromide hydrate, acmc-20alea, br2co.h2o, cobaltous bromide hydrate, dibromocobalt-water 1/1, cobalt ii bromide hexahydrate, cobalt bromide cobr2 ,hydrate 9ci | |
| MOTZAWYGLXXRSO-UHFFFAOYSA-L | |
| dibromocobalt;hydrate | |
| 16212115 | |
| 17.0 to 20.3% (Co) |
| 85017-77-2 | |
| Br2Co·xH2O | |
| MFCD00150208 | |
| Solubility in water: soluble in water | |
| O.[Co](Br)Br | |
| 218.75 | |
| 218.75 | |
| Plastic bottle |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Suspected of causing cancer.
Causes serious eye irritation.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO â Research Use Only