missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) acetate tetrahydrate, Co 24%

CAS: 6147-53-1 | C4H14CoO8 | 249.081 g/mol

Supplier:  Thermo Scientific Chemicals 012227A1

 View more versions of this product

Catalog No. AA12227A1


Only null left
Explore available promotions

Description

Description

It is used as an industrial catalyst that allow paints and varnishes to harden. Also as a precursor to various oil drying agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cobalt(II) acetate tetrahydrate
Co 24%
Red
1 kg
C4H14CoO8
3731397
Soluble in water,alcohol,dilute acids
CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Co+2]
249.081
249.08 (177.02 Anhydrous)
1.542
1.71
∽140°C-4H2O
6147-53-1
Crystals
24%
MFCD00149569
cobalt ii acetate tetrahydrate, cobalt acetate tetrahydrate, cobaltous acetate tetrahydrate, cobalt diacetate tetrahydrate, bis acetato tetraquacobalt, octan kobaltnaty czech, acetic acid, cobalt 2+ salt, tetrahydrate, octan kobaltnaty, ccris 9441
ZBYYWKJVSFHYJL-UHFFFAOYSA-L
cobalt(2+);diacetate;tetrahydrate
61372
Odorless
Hygroscopic

Chemical Identifiers

6147-53-1
249.081
ZBYYWKJVSFHYJL-UHFFFAOYSA-L
61372
CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Co+2]
C4H14CoO8
MFCD00149569
cobalt ii acetate tetrahydrate, cobalt acetate tetrahydrate, cobaltous acetate tetrahydrate, cobalt diacetate tetrahydrate, bis acetato tetraquacobalt, octan kobaltnaty czech, acetic acid, cobalt 2+ salt, tetrahydrate, octan kobaltnaty, ccris 9441
cobalt(2+);diacetate;tetrahydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.

P201-P202-P261-P271-P272-P280g-P281-P285-P302+P352-P304+P340-P308+P313-P333+P313-P342+P311-P363-P501c

H302-H317-H334-H335-H341-H350i-H360F

DOTInformation : DOT Class: 9, Packaging Group: III

EINECSNumber : 200-755-8

RTECSNumber : AG3325000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only