missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) acetate tetrahydrate, Co 24%

Catalog No. AA1222730
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1222730 250 g
AA12227A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1222730 Supplier Thermo Scientific Chemicals Supplier No. 01222730
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6147-53-1 | C4H14CoO8 | 249.081 g/mol

It is used as an industrial catalyst that allow paints and varnishes to harden. Also as a precursor to various oil drying agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6147-53-1
Molecular Formula C4H14CoO8
Molecular Weight (g/mol) 249.081
MDL Number MFCD00149569
InChI Key ZBYYWKJVSFHYJL-UHFFFAOYSA-L
Synonym cobalt ii acetate tetrahydrate, cobalt acetate tetrahydrate, cobaltous acetate tetrahydrate, cobalt diacetate tetrahydrate, bis acetato tetraquacobalt, octan kobaltnaty czech, acetic acid, cobalt 2+ salt, tetrahydrate, octan kobaltnaty, ccris 9441
PubChem CID 61372
IUPAC Name cobalt(2+);diacetate;tetrahydrate
SMILES CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Co+2]

Specifications

Melting Point ∽140°C-4H2O
Color Red
Physical Form Crystals
Quantity 250 g
Assay Percent Range 24%
Beilstein 3731397
Solubility Information Soluble in water,alcohol,dilute acids
Formula Weight 249.08 (177.02 Anhydrous)
Odor Odorless
Refractive Index 1.542
Sensitivity Hygroscopic
Density 1.71 g/cm3
Chemical Name or Material Cobalt(II) acetate tetrahydrate, Co 24%
Show More Show Less
Hazard Category H302-H317-H334-H335-H341-H350i-H360F
Hazard Statement Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
Precautionary Statement P201-P202-P261-P271-P272-P280g-P281-P285-P302+P352-P304+P340-P308+P313-P333+P313-P342+P311-P363-P501c
DOTInformation DOT Class: 9, Packaging Group: III
EINECSNumber 200-755-8
RTECSNumber AG3325000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only