missing translation for 'onlineSavingsMsg'
Learn More

Cobalt foil, 0.1mm (0.004in) thick, Puratronic™, 99.995% (metals basis)

Catalog No. p-7048778
Change view
Click to view available options
Quantity:
100 x 100 mm
25 x 25 mm
50 x 50 mm
Catalog No. Quantity
AA11455FF 25 x 25 mm
AA11455FI 50 x 50 mm
AA11455GH 100 x 100 mm
3 options

Catalog No. AA11455FF

Supplier: Thermo Scientific Chemicals 011455FF

Only null left

CAS: 7440-48-4 | Co | 58.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-48-4
Molecular Formula Co
Molecular Weight (g/mol) 58.93
MDL Number MFCD00010935
InChI Key GUTLYIVDDKVIGB-UHFFFAOYSA-N
Synonym kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
PubChem CID 104730
ChEBI CHEBI:27638
IUPAC Name cobalt
SMILES [Co]

Specifications

Quantity 25 x 25 mm
Odor Odorless
Weight 0.56g/25x25mm
Form Foil
Percent Purity 99.995%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Cobalt foil, 0.1mm (0.004 in.) thick
Hazard Category H317-H334-H350
Hazard Statement GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
EINECSNumber 231-158-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only