missing translation for 'onlineSavingsMsg'
Learn More

Cobalt foil, 0.1mm (0.004in) thick, 99.95% (metals basis)

Form: Approx.0.56g/25 x 25mm | CAS: 7440-48-4 | Co | 58.93 g/mol

Supplier:  Thermo Scientific Chemicals 042658FI

 View more versions of this product

Catalog No. AA42658FI


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Gray
7440-48-4
50 x 50 mm
99.95% (Metals basis)
Co
kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
[Co]
58.93
CHEBI:27638
Cobalt
Gray foil
Odorless
Foil
MFCD00010935
GUTLYIVDDKVIGB-UHFFFAOYSA-N
cobalt
104730

Chemical Identifiers

7440-48-4
58.93
GUTLYIVDDKVIGB-UHFFFAOYSA-N
104730
cobalt
Co
MFCD00010935
kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
CHEBI:27638
[Co]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H317

P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c

H317-H334-H350

EINECSNumber : 231-158-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only