missing translation for 'onlineSavingsMsg'
Learn More

Cobalt foil, 0.127mm (0.005in) thick, 50x50mm (2x2in), 99.9% (metals basis)

Catalog No. p-7050919
Change view
Click to view available options
Quantity:
1 Pc.
5 Pc.
Catalog No. Quantity
AA39715KG 1 Pc.
AA39715KJ 5 Pc.
2 options

Catalog No. AA39715KG

Supplier: Thermo Scientific Chemicals 039715KG

Only null left

CAS: 7440-48-4 | Co | 58.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-48-4
Molecular Formula Co
Molecular Weight (g/mol) 58.93
MDL Number MFCD00010935
InChI Key GUTLYIVDDKVIGB-UHFFFAOYSA-N
Synonym kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
PubChem CID 104730
ChEBI CHEBI:27638
IUPAC Name cobalt
SMILES [Co]

Specifications

Quantity 1 Pc.
Odor Odorless
Weight ≈2.83g/50x50mm
Form Foil
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Cobalt foil, 0.127mm (0.005 in.) thick, 50x50mm (2x2 in.)
Hazard Category H317-H334-H350
Hazard Statement GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
EINECSNumber 231-158-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only