missing translation for 'onlineSavingsMsg'
Learn More

Cobalt bis(trifluoromethylsulfonyl)imide

CAS: 207861-61-8 | C4CoF12N2O8S4 | 619.204 g/mol

$218.90 - $972.68

Chemical Identifiers

CAS 207861-61-8
Molecular Formula C4CoF12N2O8S4
Molecular Weight (g/mol) 619.204
MDL Number MFCD23380169
InChI Key HRURXKIZWNSHQB-UHFFFAOYSA-N
Synonym cobalt bis-trifluoromethanesulfonimide
PubChem CID 131875824
IUPAC Name bis(trifluoromethylsulfonyl)azanide;cobalt(2+)
SMILES C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Co+2]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4705303
View Documents
Thermo Scientific Chemicals
04705303
1 g
Each for $218.90
Only null left
 
AA4705306
View Documents
Thermo Scientific Chemicals
04705306
5 g
Each for $972.68
Only null left
 
Description

Description

Cobalt(II) Bis(trifluoromethanesulfonyl)imide is used as Surface treatment, Synthesis and catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

207861-61-8
619.204
HRURXKIZWNSHQB-UHFFFAOYSA-N
131875824
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Co+2]
C4CoF12N2O8S4
MFCD23380169
cobalt bis-trifluoromethanesulfonimide
bis(trifluoromethylsulfonyl)azanide;cobalt(2+)

Specifications

>200°C
Purple
1 g
[(CF3SO2)2N]2Co
UN3261
It is soluble in water.
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Co+2]
619.204
619.23
Cobalt bis(trifluoromethylsulfonyl)imide
207861-61-8
Solid
C4CoF12N2O8S4
MFCD23380169
cobalt bis-trifluoromethanesulfonimide
HRURXKIZWNSHQB-UHFFFAOYSA-N
bis(trifluoromethylsulfonyl)azanide;cobalt(2+)
131875824
Hygroscopic
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

P260-P264b-P271-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c

H314-H317-H335

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

RUO – Research Use Only