Learn More
Cobalt, 99.995%, (trace metal basis), Thermo Scientific Chemicals
$163.79 - $163.79
Chemical Identifiers
| CAS | 7440-48-4 |
|---|---|
| Molecular Formula | Co |
| Molecular Weight (g/mol) | 58.93 |
| MDL Number | MFCD00010935 |
| InChI Key | GUTLYIVDDKVIGB-UHFFFAOYSA-N |
| Synonym | kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride |
| PubChem CID | 104730 |
| ChEBI | CHEBI:27638 |
| IUPAC Name | cobalt |
| SMILES | [Co] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC192850050
|
Thermo Scientific Chemicals
192850050 |
5 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisChemical Identifiers
| 7440-48-4 | |
| 58.93 | |
| GUTLYIVDDKVIGB-UHFFFAOYSA-N | |
| 104730 | |
| cobalt |
| Co | |
| MFCD00010935 | |
| kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride | |
| CHEBI:27638 | |
| [Co] |
Specifications
| Pink to Violet | |
| 7440-48-4 | |
| 5 g | |
| 99.995% | |
| Co | |
| kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride | |
| [Co] | |
| 58.93 | |
| CHEBI:27638 | |
| 99.995% | |
| Cobalt |
| 1495.0°C | |
| 2870.0°C | |
| Glass bottle | |
| Powder | |
| MFCD00010935 | |
| GUTLYIVDDKVIGB-UHFFFAOYSA-N | |
| cobalt | |
| 104730 | |
| 58.93 | |
| Trace Metal Basis |
Safety and Handling
GHS P Statement Flammable solid. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Harmful if swallowed.
GHS P Statement Use personal protective equipment as required. Obtain special instructions before use. IF ON SKIN: Wash with plenty of soap and water. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Danger
RUO – Research Use Only